Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

β-Amyrin, ≥98.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014617 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4617 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4617 Supplier MilliporeSigma Supelco Supplier No. 0923610MGF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-Amyrins are pentacyclic triterpenes and a large class of secondary metabolites, isolated from various plants. It exhibits gastroprotective, antipruritic and hepatoprotective activities.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 559-70-6
Linear Formula C30H50 O
MDL Number MFCD00017381
Synonym beta-Amyrenol; Olean-12-en-3 beta-ol
Type Analytical Standard
Quantity 10 mg
Formula Weight 426.72
Percent Purity ≥98.5% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.