Learn More
Medchemexpress LLC α-Amyrin acetate | 863-76-3 | 5 MG
SDP
Catalog No. 5000421540
Encompass_Preferred
α-Amyrin acetate is a pentacyclic triterpenoid compound with oral activity, used in research related to cancer and metabolic diseases such as breast cancer and diabetes.
  • Inhibits cancer cell proliferation and induces apoptosis
  • Reduces blood glucose levels
  • Purity of 99.63%
  • White to off-white solid appearance
Catalog No. 50-004-21540 Supplier Medchemexpress LLC Supplier No. HYN28425MG
α-Amyrin acetate is a pentacyclic triterpenoid compound with oral activity, used in research related to cancer and metabolic diseases such as breast cancer and diabetes.
  • Inhibits cancer cell proliferation and induces apoptosis
  • Reduces blood glucose levels
  • Purity of 99.63%
  • White to off-white solid appearance