Learn More
Cayman Chemical Amyrn 10mg
SDP
Catalog No. 502983676
Encompass_Preferred
A selective RORa and RORY agonist that stimulates ROR transcriptional activity in HEK293 cell reporter assays at concentrations as low as 2 UM without effect at the related LXR and FXR receptors stabilizes p53 and induces apoptosis in HepG2 cancer cells
Catalog No. 50-298-3676 Supplier Cayman Chemical Supplier No. 20949 10MG
A selective RORa and RORY agonist that stimulates ROR transcriptional activity in HEK293 cell reporter assays at concentrations as low as 2 UM without effect at the related LXR and FXR receptors stabilizes p53 and induces apoptosis in HepG2 cancer cells