Learn More
Cederlane Amytracker 520, green fluorescent optotracer, 200 μL, 1 mg/mL, non-toxic, crosses blood-brain barrier, excitation 460 nm, emission 520 nm
SDP
Catalog No. NC2306207
Encompass
Amytracker 520, green fluorescent optotracer, 200 μL, 1 mg/mL, non-toxic, crosses blood-brain barrier, excitation 460 nm, emission 520 nm
Catalog No. NC2306207 Supplier Cederlane Supplier No. A520200
Amytracker 520, green fluorescent optotracer, 200 μL, 1 mg/mL, non-toxic, crosses blood-brain barrier, excitation 460 nm, emission 520 nm