Learn More
Cederlane AMYTRACKER 680 Aqueous, red fluorescent optotracer, 1 mg/ml solution, for labeling protein aggregates, non-toxic
SDP
Catalog No. NC3104887
Encompass
AMYTRACKER 680 Aqueous, red fluorescent optotracer, 1 mg/ml solution, for labeling protein aggregates, non-toxic
Catalog No. NC3104887 Supplier Cederlane Supplier No. A680A100
AMYTRACKER 680 Aqueous, red fluorescent optotracer, 1 mg/ml solution, for labeling protein aggregates, non-toxic