Learn More
Medchemexpress LLC AMZ30 | 1313613-09-0 | 99.2% | 1 ML
SDP
Catalog No. 5000340609
Encompass_Preferred
AMZ30 is a selective protein phosphatase methylesterase-1 (PME-1) inhibitor, demonstrating significant selectivity over other serine hydrolases. It reduces the demethylated form of PP2A in living cells, attenuates muscle cell differentiation, and enhances the resistance of glioblastoma cells to oxidative stress, making it suitable for glioblastoma research.
  • Selective protein phosphatase methylesterase-1 (PME-1) inhibitor
  • >100-fold selectivity compared to other serine hydrolases
  • Reduces demethylated form of PP2A in living cells
  • Attenuates muscle cell differentiation
  • Increases resistance to t-BHP-induced oxidative stress in glioblastoma cells
  • Can be used for glioblastoma research
Catalog No. 50-003-40609 Supplier Medchemexpress LLC Supplier No. HY1283310MM/1ML
AMZ30 is a selective protein phosphatase methylesterase-1 (PME-1) inhibitor, demonstrating significant selectivity over other serine hydrolases. It reduces the demethylated form of PP2A in living cells, attenuates muscle cell differentiation, and enhances the resistance of glioblastoma cells to oxidative stress, making it suitable for glioblastoma research.
  • Selective protein phosphatase methylesterase-1 (PME-1) inhibitor
  • >100-fold selectivity compared to other serine hydrolases
  • Reduces demethylated form of PP2A in living cells
  • Attenuates muscle cell differentiation
  • Increases resistance to t-BHP-induced oxidative stress in glioblastoma cells
  • Can be used for glioblastoma research