Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC An-12-h5 | 1193340-31-6 | 98.0% | 1 ML
SDP

Catalog No. 5000233685
Encompass_Preferred

AN-12-H5 is a small-molecule antienterovirus research reagent (CAS 1193340-31-6) supplied as solid material and as a ready-to-use 10 mM solution in DMSO. It is reported to inhibit replication of poliovirus and enterovirus 71 and is provided with purity specification and SDS documentation.

  • Provided as 10 mM, 1 mL solution in DMSO or as solid in multiple sizes.
  • High purity suitable for biological assays (98.0%).
  • Molecular weight 513.65; formula C24H23N3O4S3.
  • Comes with safety data sheet (SDS) and documentation.
  • Intended for research use in enterovirus replication studies.

Catalog No. 50-002-33685 Supplier Medchemexpress LLC Supplier No. HY12090010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AN-12-H5 is a small-molecule antienterovirus research reagent (CAS 1193340-31-6) supplied as solid material and as a ready-to-use 10 mM solution in DMSO. It is reported to inhibit replication of poliovirus and enterovirus 71 and is provided with purity specification and SDS documentation.

  • Provided as 10 mM, 1 mL solution in DMSO or as solid in multiple sizes.
  • High purity suitable for biological assays (98.0%).
  • Molecular weight 513.65; formula C24H23N3O4S3.
  • Comes with safety data sheet (SDS) and documentation.
  • Intended for research use in enterovirus replication studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.