Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC An-12-h5 | 1193340-31-6 | 98.0% | 100 MG
SDP

Catalog No. 5000232988
Encompass_Preferred

An antienterovirus compound that inhibits replication of poliovirus (PV) and enterovirus 71 (EV71). Supplied as a high-purity solid research reagent, it is suitable for in vitro assays and can be formulated for in vivo dosing following the provided protocols.

  • Targets replication of poliovirus and enterovirus 71.
  • High purity: 98.0%.
  • Molecular weight 513.65 g·mol⁻¹; formula C24H23N3O4S3.
  • Soluble in DMSO (100 mg/mL) for in vitro applications.
  • Available in solid packages including 100 mg and as a 10 mM solution in DMSO.
  • Storage guidance provided for powder and solutions to preserve stability.

Catalog No. 50-002-32988 Supplier Medchemexpress LLC Supplier No. HY120900100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

An antienterovirus compound that inhibits replication of poliovirus (PV) and enterovirus 71 (EV71). Supplied as a high-purity solid research reagent, it is suitable for in vitro assays and can be formulated for in vivo dosing following the provided protocols.

  • Targets replication of poliovirus and enterovirus 71.
  • High purity: 98.0%.
  • Molecular weight 513.65 g·mol⁻¹; formula C24H23N3O4S3.
  • Soluble in DMSO (100 mg/mL) for in vitro applications.
  • Available in solid packages including 100 mg and as a 10 mM solution in DMSO.
  • Storage guidance provided for powder and solutions to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.