Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC An-12-h5 | 1193340-31-6 | 98.0% | 5 MG
SDP

Catalog No. 5000234719
Encompass_Preferred

AN-12-H5 is a small-molecule antienterovirus research compound that inhibits replication of poliovirus (PV) and enterovirus 71 (EV71). It is supplied for in vitro research use and includes recommended solubility and storage conditions.

  • Antienterovirus compound targeting replication of PV and EV71.
  • Purity 98.0%.
  • Molecular formula C24H23N3O4S3 and molecular weight 513.65 g/mol.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical applications.

Catalog No. 50-002-34719 Supplier Medchemexpress LLC Supplier No. HY1209005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AN-12-H5 is a small-molecule antienterovirus research compound that inhibits replication of poliovirus (PV) and enterovirus 71 (EV71). It is supplied for in vitro research use and includes recommended solubility and storage conditions.

  • Antienterovirus compound targeting replication of PV and EV71.
  • Purity 98.0%.
  • Molecular formula C24H23N3O4S3 and molecular weight 513.65 g/mol.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.