Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AN-12-H5 intermediate-1 | 254882-14-9 | 99.7% | 259.30 | 1 ML
SDP

Catalog No. 5000400713
Encompass_Preferred

AN-12-H5 intermediate-1 is an intermediate of the viral inhibitor AN-12-H5 and can be used to synthesize Antibody-Drug Conjugates (ADCs). It is for research use only. AN-12-H5, an antienterovirus compound, targets the replication processes of PV and enterovirus 71 (EV71).

  • Intermediate for viral inhibitor
  • Used to synthesize antibody-drug conjugates (ADCs)
  • Targets replication processes of PV and enterovirus 71 (EV71)
  • Purity of 99.72%
  • Molecular weight of 259.30
  • Viscous liquid, colorless to light yellow appearance
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-004-00713 Supplier Medchemexpress LLC Supplier No. HY22055B10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

AN-12-H5 intermediate-1 is an intermediate of the viral inhibitor AN-12-H5 and can be used to synthesize Antibody-Drug Conjugates (ADCs). It is for research use only. AN-12-H5, an antienterovirus compound, targets the replication processes of PV and enterovirus 71 (EV71).

  • Intermediate for viral inhibitor
  • Used to synthesize antibody-drug conjugates (ADCs)
  • Targets replication processes of PV and enterovirus 71 (EV71)
  • Purity of 99.72%
  • Molecular weight of 259.30
  • Viscous liquid, colorless to light yellow appearance
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.