Learn More
Medchemexpress LLC AN-12-H5 intermediate-1 | 254882-14-9 | 99.7% | 259.30 | 100 MG
SDP
Catalog No. 5000401163
Encompass_Preferred
AN-12-H5 intermediate-1 is an intermediate of the viral inhibitor AN-12-H5 (HY-120900) and can be used to synthesize Antibody-Drug Conjugates (ADCs). It shows no inhibitory effect on PI4KB activity and only moderate inhibitory effects on PI 3-kinase activity.
  • Designed for synthesizing antibody-drug conjugates (ADCs)
  • Intermediate for the viral inhibitor AN-12-H5
  • Shows moderate inhibitory effects on PI 3-kinase activity
Catalog No. 50-004-01163 Supplier Medchemexpress LLC Supplier No. HY22055B100MG
AN-12-H5 intermediate-1 is an intermediate of the viral inhibitor AN-12-H5 (HY-120900) and can be used to synthesize Antibody-Drug Conjugates (ADCs). It shows no inhibitory effect on PI4KB activity and only moderate inhibitory effects on PI 3-kinase activity.
  • Designed for synthesizing antibody-drug conjugates (ADCs)
  • Intermediate for the viral inhibitor AN-12-H5
  • Shows moderate inhibitory effects on PI 3-kinase activity