Learn More
Medchemexpress LLC AN-12-H5 intermediate-1 | 254882-14-9 | 99.7% | 259.30 | 50 MG
SDP
Catalog No. 5000400715
Encompass_Preferred
AN-12-H5 intermediate-1 is a compound primarily used in the synthesis of the viral inhibitor AN-12-H5 and for developing Antibody-Drug Conjugates (ADCs). The viral inhibitor AN-12-H5 specifically targets the replication of Poliovirus (PV) and Enterovirus 71 (EV71).
  • Precursor for viral inhibitor synthesis.
  • Applicable in antibody-drug conjugate (ADC) development.
  • Exhibits high purity for research use.
Catalog No. 50-004-00715 Supplier Medchemexpress LLC Supplier No. HY22055B50MG
AN-12-H5 intermediate-1 is a compound primarily used in the synthesis of the viral inhibitor AN-12-H5 and for developing Antibody-Drug Conjugates (ADCs). The viral inhibitor AN-12-H5 specifically targets the replication of Poliovirus (PV) and Enterovirus 71 (EV71).
  • Precursor for viral inhibitor synthesis.
  • Applicable in antibody-drug conjugate (ADC) development.
  • Exhibits high purity for research use.