Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC An-12-H5 intermediate-2 | 756486-14-3 | 97.0% | 257.29 g/mol | C12H19NO5 | 5 G
SDP

Catalog No. 5000210902
Encompass_Preferred

A synthetic intermediate used in the preparation of antiviral inhibitors and as a building block for antibody-drug conjugate (ADC) synthesis. Supplied for research and synthetic applications in multiple laboratory pack sizes.

  • Used to synthesize antiviral inhibitors and ADCs.
  • Reported purity 97.0%.
  • CAS number 756486-14-3.
  • Chemical formula C12H19NO5, molecular weight 257.29 g/mol.
  • Available in multiple pack sizes including 5 g for laboratory use.

Catalog No. 50-002-10902 Supplier Medchemexpress LLC Supplier No. HYW0474965G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A synthetic intermediate used in the preparation of antiviral inhibitors and as a building block for antibody-drug conjugate (ADC) synthesis. Supplied for research and synthetic applications in multiple laboratory pack sizes.

  • Used to synthesize antiviral inhibitors and ADCs.
  • Reported purity 97.0%.
  • CAS number 756486-14-3.
  • Chemical formula C12H19NO5, molecular weight 257.29 g/mol.
  • Available in multiple pack sizes including 5 g for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.