Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC An-12-H5 intermediate-3 | 851314-03-9 | 95.8% | 330.18 g/mol | C13H16BrNO4 | 500 MG
SDP

Catalog No. 5000281482
Encompass_Preferred

AN-12-H5 intermediate-3 is a chemical intermediate used to synthesize the antiviral inhibitor AN-12-H5 and as a building block for antibody-drug conjugate (ADC) development. The material is supplied for research use and is characterized by reported purity, molecular identifiers, and supplier storage recommendations.

  • High reported purity of 95.8%.
  • Molecular formula C13H16BrNO4 and molecular weight 330.18 g/mol.
  • Suitable as a building block for antiviral compound synthesis and ADC conjugation.
  • Available in research pack sizes including 500 mg.
  • Recommended storage: powder at -20°C for long-term, 4°C for short-term.

Catalog No. 50-002-81482 Supplier Medchemexpress LLC Supplier No. HYW451820500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AN-12-H5 intermediate-3 is a chemical intermediate used to synthesize the antiviral inhibitor AN-12-H5 and as a building block for antibody-drug conjugate (ADC) development. The material is supplied for research use and is characterized by reported purity, molecular identifiers, and supplier storage recommendations.

  • High reported purity of 95.8%.
  • Molecular formula C13H16BrNO4 and molecular weight 330.18 g/mol.
  • Suitable as a building block for antiviral compound synthesis and ADC conjugation.
  • Available in research pack sizes including 500 mg.
  • Recommended storage: powder at -20°C for long-term, 4°C for short-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.