Learn More
Medchemexpress LLC HY-10978 100mg Medchemexpress, AN-2728 Crisaborole CAS:906673-24-3 Purity:98%
SDP
Catalog No. 501637456
Encompass_Preferred
Medchemexpress,HY-10978 100mg AN-2728 Crisaborole CAS:906673-24-3 Formula:C14H10BNO3 AN-2728 is a potent inhibitor of PDE4 and cytokine release; inhibit PDE4 with an IC50 of 0.49 μM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7456 Supplier Medchemexpress LLC Supplier No. HY10978100MG
Medchemexpress,HY-10978 100mg AN-2728 Crisaborole CAS:906673-24-3 Formula:C14H10BNO3 AN-2728 is a potent inhibitor of PDE4 and cytokine release; inhibit PDE4 with an IC50 of 0.49 μM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.