Learn More
Medchemexpress LLC AN-3485 | 1213827-99-6 | 97.4% | 289.52 g/mol | C14H13BClNO3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1832510MG
Encompass_Preferred
AN-3485 is a benzoxaborole analogue that acts as an orally active antagonist of Toll-like receptor signaling. It inhibits TLR-mediated inflammatory cytokine secretion, exhibits significant anti-inflammatory activity, and has reported anti-tumor effects. Supplied for research use only and provided as a solid or as a 10 mM solution in DMSO; follow supplier storage recommendations.
Catalog No. 50-002-98698