Learn More
Medchemexpress LLC HY-18325 5mg Medchemexpress, AN-3485 CAS:1213827-99-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY183255MG
Encompass_Preferred
Medchemexpress, HY-18325 5mg AN-3485 CAS:1213827-99-6 AN-3485 is a benzoxaborole analog, Toll-Like Receptor(TLR) inhibitor with IC50 values ranging from 18 to 580 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2037