Learn More
Bachem Sauvagine Trifluoroacetate
SDP
Supplier:  Bachem 4027592.0500
Encompass_Preferred
Sauvagine Trifluoroacetate, Pyr-Gly-Pro-Pro-Ile-Ser-Ile-Asp-Leu-Ser-Leu-Glu-Leu-Leu-Arg-Lys-Met-Ile-Glu-Ile-Glu-Lys-Gln-Glu-Lys-Glu-Lys-Gln-Gln-Ala-Ala-Asn-Asn-Arg-Leu-Leu-Leu-Asp-Thr-Ile-NH2, 74434-59-6, C202H346N56O63S, Mr 4599.38, 0.5mg. Sauvagine, frog. An active polypeptide isolated from the skin of the neotropical frog Phyllomedusa sauvagei. It exhibits a number of pharmacological actions on diuresis, the cardiovascular system and the endocrine glands.
Catalog No. 50-194-6794