Learn More
Biosurface Technologies Corp Anaerobic CDC Biofilm Reactor with Stir/Hot Plate, Mid- to High-Shear Laboratory Biofilm Growth Device, BioSurface Technologies Corporation.
Catalog No. NC3662406
Encompass
"The CDC Biofilm Reactor® consists of eight (8) polypropylene coupon holder rods suspended from a UHMW-polyethylene ported lid. Each coupon holder rod can accommodate three (3) 1/2-inch (12.7 mm) diameter coupons, for a total of twenty-four(24) coupons. The lid (with coupon holders and coupons) is mounted in a 1-liter glass vessel with side-arm discharge port. A liquid growth media or biocide is circulated through the vessel while mixing and shear is generated by a magnetic stir bar and vane rotated by a magnetic stir plate.Sampling of the coupons is conducted by aseptically removing individual coupon holder rods with accompanying coupons (3). The coupon holder (or blank) rod is replaced in the lid after sampling to allow the time-course experiment to continue. The coupon removed from the reactor vessel is then used for treatment evaluations, scraped or vortexed to collect the biofilm sample for further study.
Catalog No. NC3662406 Supplier Biosurface Technologies Corp Supplier No. CBR 903DH
"The CDC Biofilm Reactor® consists of eight (8) polypropylene coupon holder rods suspended from a UHMW-polyethylene ported lid. Each coupon holder rod can accommodate three (3) 1/2-inch (12.7 mm) diameter coupons, for a total of twenty-four(24) coupons. The lid (with coupon holders and coupons) is mounted in a 1-liter glass vessel with side-arm discharge port. A liquid growth media or biocide is circulated through the vessel while mixing and shear is generated by a magnetic stir bar and vane rotated by a magnetic stir plate.Sampling of the coupons is conducted by aseptically removing individual coupon holder rods with accompanying coupons (3). The coupon holder (or blank) rod is replaced in the lid after sampling to allow the time-course experiment to continue. The coupon removed from the reactor vessel is then used for treatment evaluations, scraped or vortexed to collect the biofilm sample for further study.