Learn More
Bellco Glass, Inc. Anaerobic Bottle, 100mL with 45mm S/C
SDP
Catalog No. 501945350
Encompass_Preferred
The Anaerobic Culture Bottle is designed for the growth of anaerobic bacteria and maintaining anaerobic conditions. The bottle features a 45 GL screw cap with silicone o-ring and blue butyl rubber stopper. This allows for easy access of contents and eliminates the use of the traditional aluminum seal. Butyl rubber stopper allows for multiple punctures with a needle. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassDimensions: 56mm OD X 100mm OAHPolypropylene Screw Cap With Silicone Gasket And Butyl Rubber StopperComponents Are Fully Autoclavable @ 121° CCase Quantity: 12
Catalog No. 50-194-5350 Supplier Bellco Glass, Inc. Supplier No. 204850100
The Anaerobic Culture Bottle is designed for the growth of anaerobic bacteria and maintaining anaerobic conditions. The bottle features a 45 GL screw cap with silicone o-ring and blue butyl rubber stopper. This allows for easy access of contents and eliminates the use of the traditional aluminum seal. Butyl rubber stopper allows for multiple punctures with a needle. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassDimensions: 56mm OD X 100mm OAHPolypropylene Screw Cap With Silicone Gasket And Butyl Rubber StopperComponents Are Fully Autoclavable @ 121° CCase Quantity: 12