Learn More
Bellco Glass, Inc. Anaerobic Bottle, 250mL with 45mm S/C
SDP
Catalog No. 501945351
Encompass_Preferred
The anaerobic culture bottle is designed for the growth of anaerobic bacteria and maintaining anaerobic conditions. The bottle features a 45mm screw cap with silicone o-ringand blue butyl rubber stopper. This allows for easy access of contents and eliminates the use of the traditional aluminum seal. Butyl rubber stopper allows for multiple punctures with a needle. Bottle is Borosilicate glass and cap is polypropylene. All components are autoclavable. Polypropylene screw cap with silicone gasket and butyl rubber stopper. Components are fully autoclavable @ 121°C.. 12 per case. MADE IN THE USA.
Catalog No. 50-194-5351 Supplier Bellco Glass, Inc. Supplier No. 204850250
The anaerobic culture bottle is designed for the growth of anaerobic bacteria and maintaining anaerobic conditions. The bottle features a 45mm screw cap with silicone o-ringand blue butyl rubber stopper. This allows for easy access of contents and eliminates the use of the traditional aluminum seal. Butyl rubber stopper allows for multiple punctures with a needle. Bottle is Borosilicate glass and cap is polypropylene. All components are autoclavable. Polypropylene screw cap with silicone gasket and butyl rubber stopper. Components are fully autoclavable @ 121°C.. 12 per case. MADE IN THE USA.