Learn More
Bellco Glass, Inc. Anaerobic Media Bottle, 1000mL
SDP
Catalog No. 501945344
Encompass_Preferred
Type 1 class a Borosilicate glass that are designed for maintaining anaerobic conditions. Useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development. MADE IN THE USA.Specifications: Size: 1l Dimensions: 100mm OD x 215mm OAH. Neck Dimensions: 19.7-20.2mm OD, 13.3-13.6mm ID. Grad. Range: 100mL-900mL, every 100mL. Neck Finish: #20 Alum Seal Neck. Supply with #20 Septum Stopper & Alum Seal.
Catalog No. 50-194-5344 Supplier Bellco Glass, Inc. Supplier No. 204801000
Type 1 class a Borosilicate glass that are designed for maintaining anaerobic conditions. Useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development. MADE IN THE USA.Specifications: Size: 1l Dimensions: 100mm OD x 215mm OAH. Neck Dimensions: 19.7-20.2mm OD, 13.3-13.6mm ID. Grad. Range: 100mL-900mL, every 100mL. Neck Finish: #20 Alum Seal Neck. Supply with #20 Septum Stopper & Alum Seal.