Learn More
Bellco Glass, Inc. Anaerobic Media Bottle, 100mL
SDP
Catalog No. 501945340
Encompass_Preferred
Type 1 class a Borosilicate glass that are designed for maintaining anaerobic conditions. Useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development. MADE IN THE USA.Specifications: Size: 100ml Neck Finish: #20 Alum Seal Neck, Dimensions: 56mm OD x 100mm OAH, Neck Dimensions: 19.7-20.2mm OD, 13.3-13.6mm ID, Grad. Range: 20mL-80mL, every 20mL, Supply with #20 Septum Stopper & Alum Seal. 12 per case.
Catalog No. 50-194-5340 Supplier Bellco Glass, Inc. Supplier No. 204800100
Type 1 class a Borosilicate glass that are designed for maintaining anaerobic conditions. Useful for studies involving methane-producing bacteria, cell culture, drug discovery, and research and development. MADE IN THE USA.Specifications: Size: 100ml Neck Finish: #20 Alum Seal Neck, Dimensions: 56mm OD x 100mm OAH, Neck Dimensions: 19.7-20.2mm OD, 13.3-13.6mm ID, Grad. Range: 20mL-80mL, every 20mL, Supply with #20 Septum Stopper & Alum Seal. 12 per case.