Learn More
Bellco Glass, Inc. Anaerobic Tube Only,18x150mm
SDP
Catalog No. 501945341
Encompass_Preferred
Gas tight tube for maintianing anaerobic conditions. Useful for studies invpolving methane-producing bacteria. Gas impermeable, blue butyl rubber septum type stopper. Manufactured fro type 1 borosilicate glass. With alum seal neck. MATERIAL: NCV TUBING (N51 A). 100 per case. MADE IN THE USA.Dimensions: 18mm od x 150mm longWall thickness: 1.10mmNeck ID: 13.30 - 13.60mmFor septum stoppers: 2048-11800For alum seals : 2048-11020For seal crimper: 2048-10020
Catalog No. 50-194-5341 Supplier Bellco Glass, Inc. Supplier No. 204800150
Gas tight tube for maintianing anaerobic conditions. Useful for studies invpolving methane-producing bacteria. Gas impermeable, blue butyl rubber septum type stopper. Manufactured fro type 1 borosilicate glass. With alum seal neck. MATERIAL: NCV TUBING (N51 A). 100 per case. MADE IN THE USA.Dimensions: 18mm od x 150mm longWall thickness: 1.10mmNeck ID: 13.30 - 13.60mmFor septum stoppers: 2048-11800For alum seals : 2048-11020For seal crimper: 2048-10020