Learn More
Bellco Glass, Inc. Anaerobic Tube & Stopper 18x150mm
SDP
Catalog No. 501945348
Encompass_Preferred
Gas tight tube designed for maintaining anaerobic conditions. Useful for studies involving methane-producing bacteria. Includes gas impermeable blue butyl rubber septum type stopper. Tube is made of type 1 class Borosilicate glass. Size 18x150mm. Includes Anaerobic Tube Only,18X150mm and Septum Stopper, 20mm. MADE IN THE USA.
Catalog No. 50-194-5348 Supplier Bellco Glass, Inc. Supplier No. 204818150
Gas tight tube designed for maintaining anaerobic conditions. Useful for studies involving methane-producing bacteria. Includes gas impermeable blue butyl rubber septum type stopper. Tube is made of type 1 class Borosilicate glass. Size 18x150mm. Includes Anaerobic Tube Only,18X150mm and Septum Stopper, 20mm. MADE IN THE USA.