Learn More
Medchemexpress LLC AN0128 | 872044-70-7 | 98.95% | 400.06 | 10 MG
SDP
Catalog No. 5000345387
Encompass_Preferred
AN0128 is a boron-containing antibacterial and anti-inflammatory agent. It has demonstrated efficacy against *S. aureus*, *S. epidermidis*, *P. acnes*, and *B. subtilis* with minimum inhibitory concentration (MIC) values of 1, 0.5, 0.3, and 1 μg/mL, respectively. This product is intended for research in periodontal and cutaneous diseases.
  • Acts as an antibacterial agent.
  • Functions as an anti-inflammatory agent.
  • Inhibits the release of pro-inflammatory cytokines (TNF-α, IL-1β) in human peripheral blood mononuclear cells (PBMCs) at 10 μM.
  • Reduces inflammatory infiltrate and bone loss in experimental periodontitis.
  • Shows greater effectiveness than a common toothpaste in reducing bone loss and increasing bone volume in animal models of periodontitis.
  • Available in solid form, white to off-white in color.
Catalog No. 50-003-45387 Supplier Medchemexpress LLC Supplier No. HY1097910MG
AN0128 is a boron-containing antibacterial and anti-inflammatory agent. It has demonstrated efficacy against *S. aureus*, *S. epidermidis*, *P. acnes*, and *B. subtilis* with minimum inhibitory concentration (MIC) values of 1, 0.5, 0.3, and 1 μg/mL, respectively. This product is intended for research in periodontal and cutaneous diseases.
  • Acts as an antibacterial agent.
  • Functions as an anti-inflammatory agent.
  • Inhibits the release of pro-inflammatory cytokines (TNF-α, IL-1β) in human peripheral blood mononuclear cells (PBMCs) at 10 μM.
  • Reduces inflammatory infiltrate and bone loss in experimental periodontitis.
  • Shows greater effectiveness than a common toothpaste in reducing bone loss and increasing bone volume in animal models of periodontitis.
  • Available in solid form, white to off-white in color.