Learn More
Medchemexpress LLC AN0128 | 872044-70-7 | 98.95% | 400.06 | 50 MG
SDP
Catalog No. 5000346558
Encompass_Preferred
AN0128 is a boron-containing antibacterial and anti-inflammatory agent. It is effective against various bacteria including *S. aureus*, *S. epidermidis*, *P. acnes*, and *B. subtilis*. This compound can be utilized in the research of periodontal disease and cutaneous diseases, demonstrating strong inhibition of pro-inflammatory cytokines *in vitro* and reduction of inflammatory infiltrate and bone loss *in vivo*.
  • Boron-containing antibacterial and anti-inflammatory agent
  • Effective against *S. aureus*, *S. epidermidis*, *P. acnes*, and *B. subtilis*
  • Utilized for research on periodontal disease and cutaneous diseases
  • Inhibits release of pro-inflammatory cytokines (TNF-α, IL-1β) in PBMCs
  • Reduces inflammatory infiltrate and bone loss *in vivo*
  • Increases bone area and bone volume in experimental periodontitis rat model
  • Decreases bone loss and inflammation in experimental periodontitis rat model
Catalog No. 50-003-46558 Supplier Medchemexpress LLC Supplier No. HY1097950MG
AN0128 is a boron-containing antibacterial and anti-inflammatory agent. It is effective against various bacteria including *S. aureus*, *S. epidermidis*, *P. acnes*, and *B. subtilis*. This compound can be utilized in the research of periodontal disease and cutaneous diseases, demonstrating strong inhibition of pro-inflammatory cytokines *in vitro* and reduction of inflammatory infiltrate and bone loss *in vivo*.
  • Boron-containing antibacterial and anti-inflammatory agent
  • Effective against *S. aureus*, *S. epidermidis*, *P. acnes*, and *B. subtilis*
  • Utilized for research on periodontal disease and cutaneous diseases
  • Inhibits release of pro-inflammatory cytokines (TNF-α, IL-1β) in PBMCs
  • Reduces inflammatory infiltrate and bone loss *in vivo*
  • Increases bone area and bone volume in experimental periodontitis rat model
  • Decreases bone loss and inflammation in experimental periodontitis rat model