Learn More
HW ANDERSEN PRODUCTS OF CALIFORNIA INC AN1006 Refill Kit
SDP
Catalog No. NC2143968
Encompass
This Refill Kit comes with ampoules, liner bags, Humidichips and Dosimeters for 14 cycles. Each ampoule contains 11 g of EtO and the liner bag (sterilisation chamber) holds up to 35 Each liner bag should be vacuum sealed prior to loading into the steriliser.
Catalog No. NC2143968 Supplier HW ANDERSEN PRODUCTS OF CALIFORNIA INC Supplier No. AN1006.00
This Refill Kit comes with ampoules, liner bags, Humidichips and Dosimeters for 14 cycles. Each ampoule contains 11 g of EtO and the liner bag (sterilisation chamber) holds up to 35 Each liner bag should be vacuum sealed prior to loading into the steriliser.