Learn More
Medchemexpress LLC HY-100527 5mg Medchemexpress, AN2718 CAS:174672-06-1 Purity:>98%
SDP
Catalog No. 501967739
Encompass_Preferred
Medchemexpress, HY-100527 5mg AN2718 CAS:174672-06-1 AN2718 inhibits fungal growth by blocking protein synthesis using the oxaborole tRNA trapping (OBORT) mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7739 Supplier Medchemexpress LLC Supplier No. HY1005275MG
Medchemexpress, HY-100527 5mg AN2718 CAS:174672-06-1 AN2718 inhibits fungal growth by blocking protein synthesis using the oxaborole tRNA trapping (OBORT) mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.