Learn More
Medchemexpress LLC HY-19830 5mg Medchemexpress, AN3199 CAS:1187187-10-5 Purity:>98%
SDP
Catalog No. 502082191
Encompass_Preferred
Medchemexpress, HY-19830 5mg AN3199 CAS:1187187-10-5 AN3199 is a PDE4 inhibitor with an IC50 of 94.5 nM. AN3199 can be used for the research of inflammation-associated diseases such as asthma and chronic obstructive pulmonary disease (COPD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2191 Supplier Medchemexpress LLC Supplier No. HY198305MG
Medchemexpress, HY-19830 5mg AN3199 CAS:1187187-10-5 AN3199 is a PDE4 inhibitor with an IC50 of 94.5 nM. AN3199 can be used for the research of inflammation-associated diseases such as asthma and chronic obstructive pulmonary disease (COPD). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.