Learn More
Cayman Chemical AN-7, Purity: ≥98%, CAS: 691410-93-2, 5 mg
SDP
Catalog No. 502887929
Encompass_Preferred
A potent antioxidant lipoic acid analog; has 1.5-fold increased maximal capacity over α-lipoic acid to stimulate glucose transport into myocytes
Catalog No. 50-288-7929 Supplier Cayman Chemical Supplier No. 100062125 MG
A potent antioxidant lipoic acid analog; has 1.5-fold increased maximal capacity over α-lipoic acid to stimulate glucose transport into myocytes