Learn More
Medchemexpress LLC HY-128337 5mg Medchemexpress, AN7973 CAS:1620899-32-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1283375MG
Encompass_Preferred
Medchemexpress, HY-128337 5mg AN7973 CAS:1620899-32-2 AN7973 is the 6-carboxamide benzoxaborole, blocks intracellular parasite development and inhibits Cryptosporidium growth. AN7973 is orally active, possesses favorable safety, stability, and PK parameters, and is an exciting drug candidate for treating cryptosporidiosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1163