Learn More
TARGETMOL CHEMICALS INC ANA-12 10MG
SDP
Catalog No. 502577147
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ANA-12 is a potent and selective TrkB antagonist with anxiolytic and antidepressant activity in mice. purity: 99%
Catalog No. 50-257-7147 Supplier TARGETMOL CHEMICALS INC Supplier No. T235910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ANA-12 is a potent and selective TrkB antagonist with anxiolytic and antidepressant activity in mice. purity: 99%