Learn More
Selleck Chemical LLC ANA-12 5mg
SDP
Supplier:  Selleck Chemical LLC S77455MG
Encompass_Preferred
ANA-12 is a selective TrkB antagonist with Kd of 10 nM and 12 ?M for the high and low affinity sites, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3300