Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anabaseine-d4 1mg | 1020719-05-4 | 164.24 g/mol | C10H8D4N2 | 1 MG
SDP

Catalog No. 5000227957
Encompass_Preferred

Anabaseine-d4 is the deuterium-labeled (d4) analogue of anabaseine provided as a research-grade isotope-labeled standard for analytical and tracer applications. It is intended for non-clinical laboratory use, including mass spectrometry quantitation, metabolic tracing, and analytical method development.

  • Deuterium-labeled (d4) analogue for use as an internal standard.
  • Intended for research use only; not for human or clinical use.
  • Suitable for mass spectrometry and analytical method development.
  • Provided as a 1 mg pack for small-scale experiments and standards.
  • Chemical formula C10H8D4N2; molecular weight 164.24 g/mol.

Catalog No. 50-002-27957 Supplier Medchemexpress LLC Supplier No. HY115766S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anabaseine-d4 is the deuterium-labeled (d4) analogue of anabaseine provided as a research-grade isotope-labeled standard for analytical and tracer applications. It is intended for non-clinical laboratory use, including mass spectrometry quantitation, metabolic tracing, and analytical method development.

  • Deuterium-labeled (d4) analogue for use as an internal standard.
  • Intended for research use only; not for human or clinical use.
  • Suitable for mass spectrometry and analytical method development.
  • Provided as a 1 mg pack for small-scale experiments and standards.
  • Chemical formula C10H8D4N2; molecular weight 164.24 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.