Learn More
Medchemexpress LLC Anabaseine-d4 5mg | 1020719-05-4 | 164.24 | C10H8D4N2 | 5 MG
SDP
Catalog No. 5000227958
Encompass_Preferred
Anabaseine-d4 (CAS 1020719-05-4) is the deuterium-labeled analog of anabaseine supplied as a small-quantity research reagent for analytical and pharmacological studies. It is used as a tracer or internal standard in mass spectrometry and for investigating nicotinic acetylcholine receptor activity.
  • Deuterium-labeled analog of anabaseine.
  • Suitable as an internal standard or tracer in analytical assays.
  • Used in studies of nicotinic acetylcholine receptors.
  • Provided in small research-use quantities, typically milligram scale.
Catalog No. 50-002-27958 Supplier Medchemexpress LLC Supplier No. HY115766S5MG
Anabaseine-d4 (CAS 1020719-05-4) is the deuterium-labeled analog of anabaseine supplied as a small-quantity research reagent for analytical and pharmacological studies. It is used as a tracer or internal standard in mass spectrometry and for investigating nicotinic acetylcholine receptor activity.
  • Deuterium-labeled analog of anabaseine.
  • Suitable as an internal standard or tracer in analytical assays.
  • Used in studies of nicotinic acetylcholine receptors.
  • Provided in small research-use quantities, typically milligram scale.