Learn More
Medchemexpress LLC (±) anabasine-d4 | 1020719-08-7 | MFCD04039400 | 166.26 | C10H10D4N2 | 1 MG
SDP
Catalog No. 5000284943
Encompass_Preferred
Anabasine-d4 (racemic) is a deuterium-labeled analogue of anabasine intended for use as an isotope-labeled internal standard or tracer in analytical and quantitative methods such as NMR, GC-MS, and LC-MS. Chemical formula C10H10D4N2; molecular weight 166.26.
  • Deuterium-labeled racemate for internal standard use.
  • Compatible with NMR, GC-MS, and LC-MS quantitative analyses.
  • Supplied in milligram quantities for analytical applications.
  • Chemical formula C10H10D4N2; molecular weight 166.26.
  • Store protected from light at 4°C; in solvent, store at -80°C for long-term stability.
Catalog No. 50-002-84943 Supplier Medchemexpress LLC Supplier No. HYW6988711MG
Anabasine-d4 (racemic) is a deuterium-labeled analogue of anabasine intended for use as an isotope-labeled internal standard or tracer in analytical and quantitative methods such as NMR, GC-MS, and LC-MS. Chemical formula C10H10D4N2; molecular weight 166.26.
  • Deuterium-labeled racemate for internal standard use.
  • Compatible with NMR, GC-MS, and LC-MS quantitative analyses.
  • Supplied in milligram quantities for analytical applications.
  • Chemical formula C10H10D4N2; molecular weight 166.26.
  • Store protected from light at 4°C; in solvent, store at -80°C for long-term stability.