Learn More
Medchemexpress LLC (±) Anabasine-d4 | 1020719-08-7 | MFCD04039400 | >95.0% | 166.26 g/mol | C10H10D4N2 | 5 MG
SDP
Catalog No. 5000283925
Encompass_Preferred
(±) Anabasine-d4 is the deuterium-labeled racemate of anabasine supplied for analytical and research use as an isotope-labeled reference or internal standard. It is intended for quantitative and metabolite studies involving nicotinic acetylcholine receptor pathways. Chemical formula C10H10D4N2; molecular weight 166.26 g/mol; CAS 1020719-08-7. Solubility: DMSO ~130 mg/mL. Store protected from light; refrigerate solid, and freeze solutions for long-term storage.
Catalog No. 50-002-83925 Supplier Medchemexpress LLC Supplier No. HYW6988715MG
(±) Anabasine-d4 is the deuterium-labeled racemate of anabasine supplied for analytical and research use as an isotope-labeled reference or internal standard. It is intended for quantitative and metabolite studies involving nicotinic acetylcholine receptor pathways. Chemical formula C10H10D4N2; molecular weight 166.26 g/mol; CAS 1020719-08-7. Solubility: DMSO ~130 mg/mL. Store protected from light; refrigerate solid, and freeze solutions for long-term storage.