Learn More
Medchemexpress LLC Anabasine (racemate) | 13078-04-1 | MFCD00006370 | 162.24 g/mol | C10H14N2 | 25 MG
SDP
Catalog No. 5000209485
Encompass_Preferred
(±) Anabasine is the racemic form of anabasine that acts as an agonist of the α7 nicotinic acetylcholine receptor and has reported anti-inflammatory and insecticidal activity.
  • Racemic form (±) of anabasine.
  • α7 nAChR agonist activity, used in receptor pharmacology studies.
  • Reported anti-inflammatory and insecticidal properties.
  • Suitable for biochemical and pharmacological research into nicotinic receptors.
  • Provided as a small-mass research pack for laboratory use.
Catalog No. 50-002-09485 Supplier Medchemexpress LLC Supplier No. 25MGHYW052144
(±) Anabasine is the racemic form of anabasine that acts as an agonist of the α7 nicotinic acetylcholine receptor and has reported anti-inflammatory and insecticidal activity.
  • Racemic form (±) of anabasine.
  • α7 nAChR agonist activity, used in receptor pharmacology studies.
  • Reported anti-inflammatory and insecticidal properties.
  • Suitable for biochemical and pharmacological research into nicotinic receptors.
  • Provided as a small-mass research pack for laboratory use.