Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anabasine (racemic) | 13078-04-1 | MFCD00006370 | 162.24 g/mol | C10H14N2 | 1 ML
SDP

Catalog No. 5000209985
Encompass_Preferred

(±) Anabasine is the racemic form of anabasine, a nicotinic acetylcholine receptor agonist supplied as a research reagent. The manufacturer provides guidance for preparing DMSO stock solutions at common concentrations and recommended storage conditions for the pure compound and solutions.

  • Racemic anabasine for research use
  • CAS number: 13078-04-1
  • Molecular formula: C10H14N2
  • Molecular weight: 162.24 g/mol
  • Soluble in DMSO; stock solution preparation guidance provided
  • Storage: pure form -20°C (long-term) or 4°C (short-term); in solvent -80°C (long-term)

Catalog No. 50-002-09985 Supplier Medchemexpress LLC Supplier No. HYW05214410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±) Anabasine is the racemic form of anabasine, a nicotinic acetylcholine receptor agonist supplied as a research reagent. The manufacturer provides guidance for preparing DMSO stock solutions at common concentrations and recommended storage conditions for the pure compound and solutions.

  • Racemic anabasine for research use
  • CAS number: 13078-04-1
  • Molecular formula: C10H14N2
  • Molecular weight: 162.24 g/mol
  • Soluble in DMSO; stock solution preparation guidance provided
  • Storage: pure form -20°C (long-term) or 4°C (short-term); in solvent -80°C (long-term)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.