Learn More
Medchemexpress LLC Anabasine ((S)-Anabasine) | 494-52-0 | 99.0% | 25 MG
SDP
Catalog No. 5000413833
Encompass_Preferred
Anabasine ((S)-Anabasine) is an alkaloid found as a minor component in tobacco (Nicotiana). It is a botanical pesticide, nicotine, and acts as a full agonist of nicotinic acetylcholine receptors (nAChRs). Anabasine induces depolarization of TE671 cells endogenously expressing human fetal muscle-type nAChRs (EC50=0.7 μM).
  • Alkaloid found in tobacco
  • Botanical pesticide
  • Full agonist of nicotinic acetylcholine receptors (nAChRs)
  • Induces depolarization of TE671 cells
Catalog No. 50-004-13833 Supplier Medchemexpress LLC Supplier No. 25MGHYB1532
Anabasine ((S)-Anabasine) is an alkaloid found as a minor component in tobacco (Nicotiana). It is a botanical pesticide, nicotine, and acts as a full agonist of nicotinic acetylcholine receptors (nAChRs). Anabasine induces depolarization of TE671 cells endogenously expressing human fetal muscle-type nAChRs (EC50=0.7 μM).
  • Alkaloid found in tobacco
  • Botanical pesticide
  • Full agonist of nicotinic acetylcholine receptors (nAChRs)
  • Induces depolarization of TE671 cells