Learn More
Medchemexpress LLC Anabasine ((S)-Anabasine) | 494-52-0 | 99.0% | 5 MG
SDP
Catalog No. 5000411381
Encompass_Preferred
Anabasine ((S)-Anabasine) is an alkaloid found as a minor component in tobacco (Nicotiana). It acts as a botanical pesticide nicotine and is a full agonist of nicotinic acetylcholine receptors (nAChRs). It induces depolarization of TE671 cells that endogenously express human fetal muscle-type nAChRs (EC50=0.7 μM).
  • Found as a minor component in tobacco
  • Acts as a botanical pesticide
  • Full agonist of nicotinic acetylcholine receptors (nAChRs)
  • Induces depolarization of TE671 cells
Catalog No. 50-004-11381 Supplier Medchemexpress LLC Supplier No. HYB15325MG
Anabasine ((S)-Anabasine) is an alkaloid found as a minor component in tobacco (Nicotiana). It acts as a botanical pesticide nicotine and is a full agonist of nicotinic acetylcholine receptors (nAChRs). It induces depolarization of TE671 cells that endogenously express human fetal muscle-type nAChRs (EC50=0.7 μM).
  • Found as a minor component in tobacco
  • Acts as a botanical pesticide
  • Full agonist of nicotinic acetylcholine receptors (nAChRs)
  • Induces depolarization of TE671 cells