Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anabasine ((S)-Anabasine) | 494-52-0 | MFCD00006370 | 99.0% | 50 MG
SDP

Catalog No. 5000411691
Encompass_Preferred

Anabasine is an alkaloid found as a minor component in tobacco (Nicotiana). It is a botanical pesticide that acts as a full agonist of nicotinic acetylcholine receptors (nAChRs). Anabasine induces depolarization of TE671 cells endogenously expressing human fetal muscle-type nAChRs (EC50=0.7 μM).

  • Found in tobacco (Nicotiana)
  • Acts as a full agonist of nicotinic acetylcholine receptors
  • Induces depolarization of TE671 cells
  • Molecular weight: 162.24
  • Formula: C10H14N2
  • Appearance: Liquid (Density: 1.0455 g/cm3)
  • Colorless to light yellow liquid
  • Storage: 4°C, protect from light

Catalog No. 50-004-11691 Supplier Medchemexpress LLC Supplier No. HYB153250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anabasine is an alkaloid found as a minor component in tobacco (Nicotiana). It is a botanical pesticide that acts as a full agonist of nicotinic acetylcholine receptors (nAChRs). Anabasine induces depolarization of TE671 cells endogenously expressing human fetal muscle-type nAChRs (EC50=0.7 μM).

  • Found in tobacco (Nicotiana)
  • Acts as a full agonist of nicotinic acetylcholine receptors
  • Induces depolarization of TE671 cells
  • Molecular weight: 162.24
  • Formula: C10H14N2
  • Appearance: Liquid (Density: 1.0455 g/cm3)
  • Colorless to light yellow liquid
  • Storage: 4°C, protect from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.