Learn More
TARGETMOL CHEMICALS INC ANACARDIC ACID 25MG
SDP
Catalog No. 502579396
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Anacardic Acid (6-pentadecylsalicylic Acid) is an effective inhibitor of p300 and p300/CBP-associated factor histone acetyltranferases. It also has antimicrobial activity antibacterial activity and inhibits prostaglandin synthase lipoxygenase and tyrosinase. purity: 99%
Catalog No. 50-257-9396 Supplier TARGETMOL CHEMICALS INC Supplier No. T638925MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Anacardic Acid (6-pentadecylsalicylic Acid) is an effective inhibitor of p300 and p300/CBP-associated factor histone acetyltranferases. It also has antimicrobial activity antibacterial activity and inhibits prostaglandin synthase lipoxygenase and tyrosinase. purity: 99%