Learn More
AdipoGen Anacardic acid (5 mg)
SDP
Supplier:  AdipoGen AGCR10046M005
Encompass_Preferred
Anacardic acid (5 mg), AdipoGen Life Sciences. Chemical. CAS: 16611-84-0. Formula: C22H36O3. MW: 348.5. Potent histone acetyltransferase (HAT) inhibitor. Antibacterial. SUMOylation inhibitor. Lipoxygenase inhibitor. NF-kappaB inhibitor. Anti-tumor compound. Anti-inflammatory. Apoptosis inducer. Antiproliferative.
Catalog No. 50-149-0060