Learn More
Enzo Life Sciences Anacardic acid (5mg). CAS: 16611-84-0
SDP
Supplier:  Enzo Life Sciences ALX270381M005
Encompass_Preferred
Cell permeable salicylic acid analog that acts as a potent, non-competitive inhibitor of p300 and PCAF (p300/CBP-associated factor) histone acetyltransferase (HAT) activities (IC50~8.5µM and ~5µM, respectively). Displays a variety of other biological activities, such as antibacterial, antimicrobial, prostaglandin synthase inhibition, tyrosinase, and lipoxygenase inhibition.Inhibits SUMOylation of RanGAP1-C2 with an IC50 = 2.2µM Alternative name: 2-Hydroxy-6-pentadecylbenzoic acid, 6-Pentadecylsalicylic acid, AA. Formula: C22H36O3. MW: 348.5. MI: 14: 621. Purity: ≥95% (HPLC). Solubility: Soluble in 100% ethanol, methanol, DMSO, dichloromethane or ethylacetate. Use/Stability: Stock solutions are stable for up to 3 months when stored at -20°C. Long Term Storage: -20°C. Handling: Protect from light. Packaged under inert gas. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8525