Learn More
Apexbio Technology LLC Anacetrapib (MK-0859) 875446-37-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061609
Encompass_Preferred
Anacetrapib (MK-0859 CAS 875446-37-0) is a selective inhibitor of cholesteryl ester transfer protein (CETP) an enzyme involved in mediating the transfer of cholesteryl esters and triglycerides between lipoproteins In biochemical assays anacetrapib exhibits potent inhibition of CETP-driven lipid transfer with reported IC50 values of 16 nM for cholesteryl ester transfer and 29 nM for triglyceride transfer Metabolically it undergoes oxidation via CYP3A4 to form metabolites M1 M2 and M3 Due to its effect of significantly lowering LDL cholesterol and elevating HDL cholesterol anacetrapib is studied in research related to primary hypercholesterolemia and mixed dyslipidemia
Catalog No. 50-000-61609 Supplier Apexbio Technology LLC Supplier No. A43755.1
 Restricted Item
Add to Cart
Add to Cart
Anacetrapib (MK-0859 CAS 875446-37-0) is a selective inhibitor of cholesteryl ester transfer protein (CETP) an enzyme involved in mediating the transfer of cholesteryl esters and triglycerides between lipoproteins In biochemical assays anacetrapib exhibits potent inhibition of CETP-driven lipid transfer with reported IC50 values of 16 nM for cholesteryl ester transfer and 29 nM for triglyceride transfer Metabolically it undergoes oxidation via CYP3A4 to form metabolites M1 M2 and M3 Due to its effect of significantly lowering LDL cholesterol and elevating HDL cholesterol anacetrapib is studied in research related to primary hypercholesterolemia and mixed dyslipidemia