Learn More
Biolog Inc ATTM - Anaerobic Tissue Transport Medium, 5 Tubes/Pkg
SDP
Supplier:  Biolog Inc AS919
Encompass
This item has a minimum qty of 2 per supplier requirements.
Anaerobic Tissue Transport Medium (ATTM) is a mineral salt based semi-solid media in a wide mouth tube. Designed as a non-nutritive holding medium, it maintains viability of microorganisms through collection & transport of clinical specimens. This media is prepared, dispensed, & packaged under oxygen-free conditions. This medium is available in multiple sizes & configurations, includ. sterile surgery packs. ATTM contains buffered mineral salts in a semi-solid media, w/ sodium thioglycollate & cysteine added as reducing agents. This medium maintains the viability of most microorganisms without multiplication & allows for dilution of inhibitors present in clinical material, & meets the viability requirements of obligate anaerobes. Tubes are 25mm wide to facilitate placement & removal of larger sized samples such as tissue biopsies or feces. Resazurin is added as indicator of significant oxygen exposure to the media.
Catalog No. NC1036556