Learn More
Chemglass Life Sciences COMPLETE Anaerobic Tubes, 18 x 150mm, Supplied with 20mm Blue Butyl Rubber Stopper and Aluminum Seal
SDP
Catalog No. 501215191
Encompass_Preferred
COMPLETE Anaerobic Tubes, 18 x 150mm, Supplied with 20mm Blue Butyl Rubber Stopper and Aluminum Seal. Chemglass Life Sciences’ “Balch-type” tubes are modified versions of screw thread Hungate tubes. They feature component parts designed and engineered to maximize the effective seal between the walls of the tubes and the stoppers, providing a gas-tight seal for improved stability. The special formulation of our blue, chlorobutyl rubber stoppers are heavier than septa and more capable of withstanding repeated piercings with syringes. A visibly crimped aluminum seal is also more secure than a screw thread cap that may or may not have been tightened completely. Disposable glass tubes measure 18 x 150mm and are made from Type I, Class B borosilicate glass. Complete item consists of 100 each of disposable glass tubes, blue chlorobutyl stoppers and aluminum seals.
Catalog No. 50-121-5191 Supplier Chemglass Life Sciences Supplier No. CLS420901
COMPLETE Anaerobic Tubes, 18 x 150mm, Supplied with 20mm Blue Butyl Rubber Stopper and Aluminum Seal. Chemglass Life Sciences’ “Balch-type” tubes are modified versions of screw thread Hungate tubes. They feature component parts designed and engineered to maximize the effective seal between the walls of the tubes and the stoppers, providing a gas-tight seal for improved stability. The special formulation of our blue, chlorobutyl rubber stoppers are heavier than septa and more capable of withstanding repeated piercings with syringes. A visibly crimped aluminum seal is also more secure than a screw thread cap that may or may not have been tightened completely. Disposable glass tubes measure 18 x 150mm and are made from Type I, Class B borosilicate glass. Complete item consists of 100 each of disposable glass tubes, blue chlorobutyl stoppers and aluminum seals.