Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ AnaeroGRO™ Brucella with Hemin and Vitamin K, Anaerobe Agar
SDP

Catalog No. 23001804 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Product Type: Agar Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-001-804 Product Type: Agar Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-804 Supplier Hardy Diagnostics™ Supplier No. AG301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

AnaeroGro™ Brucella Agar with Hemin and Vitamin K is recommended for use in the primary isolation, quantitation, and partial identification of obligately anaerobic microorganisms from clinical specimens.

  • The medium is also suitable for the growth of aerobic and microaerophilic bacteria when incubated under the appropriate conditions
  • Pre-reducted culture media packaged in oxygen-free gas flushed foil pouches
  • Packaging contains an oxygen scavenger sachet
  • Packaging contains a moisture absorbing desiccant packet
  • Room temperature storage
  • Foil pouches have an easy-to-open pre-scored notch
  • Manufactured in an ISO 13485 certified facility
  • Wide variety of packaging combinations available: monoplates, biplates, and primary set-up combinations

Specifications

Appearance Opaque and medium to dark red in color
ATCC® Yes
Color Red
Content And Storage 15°C to 30°C
Country of Origin United States
Additive Hemin, Vitamin K
Form Prepared Medium
Format Plate
Manufacturing Origin United States
Media Type Agar Plate
Product Type Agar Plate
Sterility Sterile
Target Organism Class Anaerobes, Aerobic Microorganisms
Test Type Microbial Identification
For Use With (Application) Primary isolation, quantitation, and partial identification of obligately anaerobic microorganisms, growth of aerobic and microaerophilic bacteria
Incubation Time 24 to 48 hr.
Material Polystyrene
No. of Places 1
No. per Pack 1
Dimensions (Dia. x H) 100 x 15 mm
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Species Bacteroides fragilis, Porphyromonas species, Clostridium perfringens, Fusobacterium nucleatum, Peptostreptococcus
Manufacturing Quality FDA Licensed Facility
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.